Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Normal Rabbit IgG Rabbit, Polyclonal, Isotype Control, MilliporeSigma™
GSA_VA

Catalog No. 12370MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-370-MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-370-MI Supplier MilliporeSigma Supplier No. 12370
Only null left
Add to Cart
Add to Cart

Rabbit Isotype Control

Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation and Western Blotting.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, with 0.05% sodium azide before the addition of glycerol to 30%. Liquid at –20°C.
Host Species Rabbit
Purification Method Purified
Quantity 250 μg
Primary or Secondary Primary
Content And Storage Stable for 1 year at -20°C from date of receipt.
For maximum recovery of product centrifuge the original vial prior to removing the cap. Handling Recommendations: Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.