Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Normal Rabbit Serum

Catalog No. PLN5001
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PLN5001 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PLN5001 Supplier Invitrogen™ Supplier No. PLN5001
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Normal rabbit serum.

Normal rabbit serum.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Species Rabbit
Form Liquid
Product Type Rabbit Serum
Quantity 5 mL
Shipping Condition Dry Ice
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.