Learn More
Abcam Normal Rabbit Serum (Sterile), 25ML
Catalog No. 502709848
Encompass_Preferred
Sterile rabbit serum ab7487 for blocking in antibody-based applications or use as an antibody diluent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9848 Supplier Abcam Supplier No. AB748725ML
Sterile rabbit serum ab7487 for blocking in antibody-based applications or use as an antibody diluent.
The product is subject to the following: Abcam Restricted Use Statement