Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Normal Rat Serum, Invitrogen™

Catalog No. 019601
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-960-1 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-960-1 Supplier Invitrogen Supplier No. 019601
Only null left
Add to Cart
Add to Cart

Rat Control

Normal Rat Serum Control, 01-9601, from Invitrogen™. Species Reactivity: Not Applicable; Applications: Control.

Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components. Normal serum is frequently used for blocking or saturating generalized binding interactions for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods.
Host Species: Rat
Isotype: Ig Mix
Immunogen: Rat immunoglobins
Formulation: 5mL unpurified control antibody in whole serum and 0.1% sodium azide
Reconstitution with 5mL of distilled water. After reconstitution, store product in frozen aliquots.
References:
Test: Flow, Species: Ms, Reference: Constitutive Bcl-2 expression during immunoglobulin heavy chain-promoted B cell differentiation expands novel precursor B cells. (Jan 1997) Immunity, Vol: 6, 1, 23-33, Young F,Mizoguchi E,Bhan AK,Alt FW; Pubmed ID: 9052834
TRUSTED_SUSTAINABILITY

Specifications

Antigen Normal Rat Serum
Conjugate Unconjugated
Formulation whole serum with 0.1% sodium azide
Host Species Rat
Immunogen Rat immunoglobins
Quantity 5 mL
Regulatory Status RUO
Target Species None
Form Lyophilized
Isotype Ig Mix
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.