Learn More
Abcam Normal Rat Serum (sterile), 50ML
Catalog No. 502709846
Encompass_Preferred
Rat serum for blocking in antibody-based applications or use as an antibody diluent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9846 Supplier Abcam Supplier No. AB748850ML
Rat serum for blocking in antibody-based applications or use as an antibody diluent.
The product is subject to the following: Abcam Restricted Use Statement