Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Normal Saline, 100 mL
SDP

Catalog No. 50146751
Encompass_Preferred

Normal Salines isotonic solution is used as a general laboratory diluent. For non clinical use. Use as a solvent, washing agent for cells/bacteria/specimen, and buffer in aseptic techniques, plaque assays, and titer assays.

With 9 g/L sodium chloride
Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
pH: 5.0 (4.5 to 7.0)
Osmolality: 276-305 mOsM/L H2O
Concentration: 0.154 mol/L
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-146-751 Supplier Quality Biological Inc Supplier No. 114055721EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Normal Salines isotonic solution is used as a general laboratory diluent. For non clinical use. Use as a solvent, washing agent for cells/bacteria/specimen, and buffer in aseptic techniques, plaque assays, and titer assays.

With 9 g/L sodium chloride
Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
pH: 5.0 (4.5 to 7.0)
Osmolality: 276-305 mOsM/L H2O
Concentration: 0.154 mol/L
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.