Learn More
Abcam Normal Sheep Serum (sterile), 50ML
Catalog No. 502709845
Encompass_Preferred
Sheep serum for blocking in antibody-based applications or use as an antibody diluent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-9845 Supplier Abcam Supplier No. AB748950ML
Sheep serum for blocking in antibody-based applications or use as an antibody diluent.
The product is subject to the following: Abcam Restricted Use Statement