Learn More
Cayman Chemical nor-Mephedrone (hydrochloride), Purity: ≥98%, CAS: 6941-17-9, 10 mg
SDP
Catalog No. 502894485
Encompass_Preferred
An analytical reference standard categorized as a cathinone metabolite; a metabolite of mephedrone; intended for research and forensic applications
Catalog No. 50-289-4485 Supplier Cayman Chemical Supplier No. 900094010 MG
An analytical reference standard categorized as a cathinone metabolite; a metabolite of mephedrone; intended for research and forensic applications