Learn More
Medchemexpress LLC Normetanephrine-d3 hydrochloride | 1085333-97-6 | 99.9% | 222.68 | 1 MG
SDP
Catalog No. 5000453421
Encompass_Preferred
Normetanephrine-d3 (hydrochloride) is the deuterium labeled Normetanephrine hydrochloride. Normetanephrine ((±)-Normetanephrine) hydrochloride is the O-methylated metabolite of norepinephrine (NE). It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-53421 Supplier Medchemexpress LLC Supplier No. HYW008794S1MG
Normetanephrine-d3 (hydrochloride) is the deuterium labeled Normetanephrine hydrochloride. Normetanephrine ((±)-Normetanephrine) hydrochloride is the O-methylated metabolite of norepinephrine (NE). It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.