Learn More
Medchemexpress LLC Normetanephrine-d3 hydrochloride | 1085333-97-6 | 99.9% | 222.68 | 10 MG
SDP
Catalog No. 5000453420
Encompass_Preferred
Normetanephrine-d3 (hydrochloride) is the deuterium labeled Normetanephrine hydrochloride. Normetanephrine ((±)-Normetanephrine) hydrochloride is the O-methylated metabolite of norepinephrine (NE). It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-53420 Supplier Medchemexpress LLC Supplier No. HYW008794S10MG
Normetanephrine-d3 (hydrochloride) is the deuterium labeled Normetanephrine hydrochloride. Normetanephrine ((±)-Normetanephrine) hydrochloride is the O-methylated metabolite of norepinephrine (NE). It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.