Learn More
TARGETMOL CHEMICALS INC Normetanephrine hydrochloride
SDP
Catalog No. 502879959
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Normetanephrine hydrochloride (DL-Normetanephrine hydrochloride) is a metabolite of Epinephrine. It is found together with Metanephrine in urine and in certain tissues. Purity 99.16%
Catalog No. 50-287-9959 Supplier TARGETMOL CHEMICALS INC Supplier No. T49455MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Normetanephrine hydrochloride (DL-Normetanephrine hydrochloride) is a metabolite of Epinephrine. It is found together with Metanephrine in urine and in certain tissues. Purity 99.16%