Learn More
Cayman Chemical NormetnphrInhydrochlorId 100mg
SDP
Catalog No. 502917648
Encompass_Preferred
An active metabolite of norepinephrine; induces contractions in isolated rabbit aortic strips; inhibits norepinephrine uptake in isolated rat heart (IC50 = 200 µM); plasma levels of normetanephrine are increased in patients with von Hippel-Lindau disease and pheochromocytomas compared with patients with von Hippel-Lindau disease without pheochromocytomas; has been used as a marker of authentic human urine specimens
Catalog No. 50-291-7648 Supplier Cayman Chemical Supplier No. 38792100 MG
An active metabolite of norepinephrine; induces contractions in isolated rabbit aortic strips; inhibits norepinephrine uptake in isolated rat heart (IC50 = 200 µM); plasma levels of normetanephrine are increased in patients with von Hippel-Lindau disease and pheochromocytomas compared with patients with von Hippel-Lindau disease without pheochromocytomas; has been used as a marker of authentic human urine specimens