Learn More
Cayman Chemical Normetanephrine (hydrochloride), Purity: ≥95%, CAS: 1011-74-1, 50 mg
SDP
Catalog No. 502917205
Encompass_Preferred
An active metabolite of norepinephrine; induces contractions in isolated rabbit aortic strips; inhibits norepinephrine uptake in isolated rat heart (IC50 = 200 µM); plasma levels of normetanephrine are increased in patients with von Hippel-Lindau disease and pheochromocytomas compared with patients with von Hippel-Lindau disease without pheochromocytomas; has been used as a marker of authentic human urine specimens
Catalog No. 50-291-7205 Supplier Cayman Chemical Supplier No. 3879250 MG
An active metabolite of norepinephrine; induces contractions in isolated rabbit aortic strips; inhibits norepinephrine uptake in isolated rat heart (IC50 = 200 µM); plasma levels of normetanephrine are increased in patients with von Hippel-Lindau disease and pheochromocytomas compared with patients with von Hippel-Lindau disease without pheochromocytomas; has been used as a marker of authentic human urine specimens