Learn More
Invivogen Normocin™ - Antimicrobial Reagent, (10 X 1ml) 500mg
SDP
Catalog No. NC9273499
Encompass
Normocin;, An Innovative Formulation of 3 Antibiotics Active against Mycoplasma, Bacteria and Fungi, It Displays very Efficient Anti-Mycoplasma Action, and it Provided as a Red Solution at a Concentration of 50mg/mL, 10 x 1mL
Catalog No. NC9273499 Supplier Invivogen Supplier No. ANTNR1
Normocin;, An Innovative Formulation of 3 Antibiotics Active against Mycoplasma, Bacteria and Fungi, It Displays very Efficient Anti-Mycoplasma Action, and it Provided as a Red Solution at a Concentration of 50mg/mL, 10 x 1mL