Learn More
Med Vet International Normosol-R Electrolyte Injection, Nonpyrogenic Isotonic Solution, IV Fluids, 1000mL
Catalog No. 502827770
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Normosol-R is a sterile, nonpyrogenic isotonic solution of balanced electrolytes in water for injection. The solution is administered by intravenous infusion for parenteral replacement of acute losses of extracellular fluid. Normosol-R is a parenteral fluid and electrolyte replenisher. When administered intravenously, Normosol-R provides water and electrolytes for replacement of acute extracellular fluid losses without disturbing normal electrolyte relationships. Normosol-R also can be used as an adjunct to restore a decrease in circulatory volume in patients with moderate blood loss. Normosol-R is not intended to supplant transfusion of whole blood or packed red cells in the presence of uncontrolled hemorrhage or severe reductions of red cell volume.
Catalog No. 50-282-7770 Supplier Med Vet International Supplier No. RXNORMOSOLR
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Normosol-R is a sterile, nonpyrogenic isotonic solution of balanced electrolytes in water for injection. The solution is administered by intravenous infusion for parenteral replacement of acute losses of extracellular fluid. Normosol-R is a parenteral fluid and electrolyte replenisher. When administered intravenously, Normosol-R provides water and electrolytes for replacement of acute extracellular fluid losses without disturbing normal electrolyte relationships. Normosol-R also can be used as an adjunct to restore a decrease in circulatory volume in patients with moderate blood loss. Normosol-R is not intended to supplant transfusion of whole blood or packed red cells in the presence of uncontrolled hemorrhage or severe reductions of red cell volume.