Learn More
Cayman Chemical -NornIcotIn 100mg
SDP
Catalog No. 502981023
Encompass_Preferred
An active metabolite of nicotine and nAChR agonist; induces currents in Xenopus oocytes expressing human α7 nAChRs or α6 subunit-containing nAChRs (EC50s = 17 and 3.7 µM, respectively); promotes cell migration and destabilizes cell-cell junctions in EA.hy926 endothelial cells; is self-administered in rats
Catalog No. 50-298-1023 Supplier Cayman Chemical Supplier No. 20505100 MG
An active metabolite of nicotine and nAChR agonist; induces currents in Xenopus oocytes expressing human α7 nAChRs or α6 subunit-containing nAChRs (EC50s = 17 and 3.7 µM, respectively); promotes cell migration and destabilizes cell-cell junctions in EA.hy926 endothelial cells; is self-administered in rats