Learn More
Cayman Chemical (±)-Nornicotine, Purity: ≥95%, CAS: 5746-86-1, 50 mg
SDP
Catalog No. 502982474
Encompass_Preferred
An active metabolite of nicotine and nAChR agonist; induces currents in Xenopus oocytes expressing human α7 nAChRs or α6 subunit-containing nAChRs (EC50s = 17 and 3.7 µM, respectively); promotes cell migration and destabilizes cell-cell junctions in EA.hy926 endothelial cells; is self-administered in rats
Catalog No. 50-298-2474 Supplier Cayman Chemical Supplier No. 2050550 MG
An active metabolite of nicotine and nAChR agonist; induces currents in Xenopus oocytes expressing human α7 nAChRs or α6 subunit-containing nAChRs (EC50s = 17 and 3.7 µM, respectively); promotes cell migration and destabilizes cell-cell junctions in EA.hy926 endothelial cells; is self-administered in rats