Learn More
Cayman Chemical -NornIcotIn-d4 1mg
SDP
Catalog No. 502910006
Encompass_Preferred
An internal standard for the quantification of (±)-nornicotine by GC- or LC-MS
Catalog No. 50-291-0006 Supplier Cayman Chemical Supplier No. 352421 MG
An internal standard for the quantification of (±)-nornicotine by GC- or LC-MS