Learn More
Medchemexpress LLC (±)-Nornicotine-d4 | 66148-18-3 | 152.23 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW002112S10MG
Encompass_Preferred
(±)-Nornicotine-d4 is a deuterium labeled (±)-Nornicotine, which is a major metabolite of Nicotine. It acts as a partial nAChRs agonist, specifically activating receptor subtypes containing α7 and α6 subunits. This compound has been shown to disrupt β-catenin and ZO-1, induce F-actin depolymerization, and support self-administration behavior.
  • Can be used in research for atherosclerosis
  • Can be used in research for Alzheimer's disease
  • Can be used in research for schizophrenia
Catalog No. 50-004-46930