Learn More
Medchemexpress LLC (±)-Nornicotine-d4 | 66148-18-3 | 152.23 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW002112S50MG
Encompass_Preferred
(±)-Nornicotine-d4 is the deuterium labeled form of (±)-Nornicotine, a major metabolite of Nicotine. It acts as a partial nAChRs agonist, specifically targeting receptor subtypes containing α7 and α6 subunits. This compound has been shown to disrupt β-catenin and ZO-1, and induce F-actin depolymerization. Deuterium labeling can affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled form of nornicotine
  • Major metabolite of nicotine
  • Acts as a partial nAChRs agonist
  • Activates receptor subtypes containing α7 and α6 subunits
  • Disrupts β-catenin and ZO-1
  • Induces F-actin depolymerization
  • Supports self-administration behavior
  • Applicable in research for atherosclerosis, Alzheimer's disease, and schizophrenia
  • Potential to affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-47409