Learn More
Cayman Chemical nor-NOHA (acetate), Purity: ≥97%, CAS: 1140844-63-8, 5 mg
SDP
Catalog No. 502890984
Encompass_Preferred
A potent arginase inhibitor (Ki = 0.5 µM for the rat liver enzyme); ~40-fold more potent than NOHA as an inhibitor of arginase from mouse macrophages (IC50 = 10-12 µM); not a substrate for any of the 3 NOS isoforms; does not inhibit nNOS or iNOS
Catalog No. 50-289-0984 Supplier Cayman Chemical Supplier No. 100068615 MG
A potent arginase inhibitor (Ki = 0.5 µM for the rat liver enzyme); ~40-fold more potent than NOHA as an inhibitor of arginase from mouse macrophages (IC50 = 10-12 µM); not a substrate for any of the 3 NOS isoforms; does not inhibit nNOS or iNOS