Learn More
Med Vet International Norbrook Noromycin 300 La (Oxytetracycline La 300), 300mg/mL, 250mL
Catalog No. 502827771
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Noromycin 300 LA is a sterile ready-to-use solution of broad? Spectrum oxytetracycline 300 mg/mL for use in beef cattle, nonlactating dairy cattle, calves, pre-ruminating (veal) calves and swine. Oxytetracycline is effective in the treatment of a wide range of diseases caused by susceptible gram-negative and gram-positive bacteria. Observe label directions and withdrawal times. Not for use in lactating dairy animals. Adverse reactions, including injection site swelling, restlessness, ataxia, trembling, respiratory abnormalities (labored breathing), collapse and possibly death have been reported. See product labeling for full product information.
Catalog No. 50-282-7771 Supplier Med Vet International Supplier No. RXNOROMYCIN250
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Noromycin 300 LA is a sterile ready-to-use solution of broad? Spectrum oxytetracycline 300 mg/mL for use in beef cattle, nonlactating dairy cattle, calves, pre-ruminating (veal) calves and swine. Oxytetracycline is effective in the treatment of a wide range of diseases caused by susceptible gram-negative and gram-positive bacteria. Observe label directions and withdrawal times. Not for use in lactating dairy animals. Adverse reactions, including injection site swelling, restlessness, ataxia, trembling, respiratory abnormalities (labored breathing), collapse and possibly death have been reported. See product labeling for full product information.