Learn More
Bioss Norovirus
SDP
Catalog No. 5000065311
Encompass_Preferred
Norovirus Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65311 Supplier Bioss Supplier No. BSM72355M
Norovirus Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide