Learn More
Description
Positive control includes:
- Recombinant Norovirus GI.1 (ORF1-ORF2)
- Recombinant Norovirus GII.4 (ORF1-ORF2)
Negative control includes Rotavirus
Specifications
Specifications
| Biosafety Level | 1 |
| Content And Storage | 2 to 25°C |
| Format | Helix Elite |
| Manufacturing Origin | United States |
| Product Type | Helix Elite Control Reference Panels and Standards |
| For Use With (Application) | Clinical |
| Packaging Type | Plastic Box |
| Quantity | 12 Swabs |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
