Learn More
Creative Biomart Vibrio cholerae INABA 48 Classical Biotype, Heat Inactivated Antigen
Supplier:  Creative Biomart NG1349
Encompass_Preferred
The Recombinant Norovirus Group-1 Capsid, E.Coli derived, is a positive sense RNA virus with 7.5kb nucleotides, encoding a major structural protein VP1 with 50-55kDa and a VP2 protein. The full length of VP1 capsid protein is derived from the group 1 Norwalk virus. The protein is fused to a 6 His tag at N-terminal and purified by chromatography techniques.
Catalog No. 50-208-4727
 Restricted Item