Learn More
Cayman Chemical Norpropranolol (hydrochloride), Purity: ≥98%, CAS: 62618-09-1, 10 mg
SDP
Catalog No. 502911162
Encompass_Preferred
An active metabolite of propranolol; binds to β-ARs (Kd = 42 nM in turkey erythrocytes); inhibits (–)-isoproterenol-induced activation of adenylate cyclase (Ki = 175 nM)
Catalog No. 50-291-1162 Supplier Cayman Chemical Supplier No. 3601110 MG
An active metabolite of propranolol; binds to β-ARs (Kd = 42 nM in turkey erythrocytes); inhibits (–)-isoproterenol-induced activation of adenylate cyclase (Ki = 175 nM)