Learn More
Medchemexpress LLC Norquetiapine-d8 (N-desalkylquetiapine-d8) | 1189866-35-0 | 303.45 | C17H9D8N3S | 1 MG
SDP
Catalog No. 5000210475
Encompass_Preferred
Norquetiapine-d8 (N-desalkylquetiapine-d8) is a deuterium-labeled analogue of norquetiapine supplied for research use as an isotopically labeled internal standard or tracer in quantitative analytical methods. It is intended for use in pharmacological and analytical studies, including method development and validation.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Isotopic label: d8; useful as a tracer or internal standard.
  • Chemical formula C17H9D8N3S; molecular weight 303.45.
  • Available as a 1 mg vial; purity and storage per certificate of analysis.
Catalog No. 50-002-10475 Supplier Medchemexpress LLC Supplier No. HYW052508S1MG
Norquetiapine-d8 (N-desalkylquetiapine-d8) is a deuterium-labeled analogue of norquetiapine supplied for research use as an isotopically labeled internal standard or tracer in quantitative analytical methods. It is intended for use in pharmacological and analytical studies, including method development and validation.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Isotopic label: d8; useful as a tracer or internal standard.
  • Chemical formula C17H9D8N3S; molecular weight 303.45.
  • Available as a 1 mg vial; purity and storage per certificate of analysis.