Learn More
Norell Inc NORELL OPTIMIZER INSERT FOR ADAPTING 4mm TUBE TO 5mm BRUKER SPINNER - U.S. PATENT NO. 7,728,593
SDP
Catalog No. NC0336652
Encompass_Preferred
NORELL OPTIMIZER INSERT FOR ADAPTING 4mm TUBE TO 5mm BRUKER SPINNER - U.S. PATENT NO. 7,728,593
Catalog No. NC0336652 Supplier Norell Inc Supplier No. 5X4INSB
NORELL OPTIMIZER INSERT FOR ADAPTING 4mm TUBE TO 5mm BRUKER SPINNER - U.S. PATENT NO. 7,728,593