Learn More
Cayman Chemical Norsufentanil-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1204688-16-3, 1 mg
SDP
Catalog No. 502895037
Encompass_Preferred
An internal standard for the quantification of norsufentanil by GC- or LC-MS
Catalog No. 50-289-5037 Supplier Cayman Chemical Supplier No. 90011431 MG
An internal standard for the quantification of norsufentanil by GC- or LC-MS