Learn More
Cayman Chemical NorsufentnIl-d3 500UG
SDP
Catalog No. 502895038
Encompass_Preferred
An internal standard for the quantification of norsufentanil by GC- or LC-MS
Catalog No. 50-289-5038 Supplier Cayman Chemical Supplier No. 9001143500 UG
An internal standard for the quantification of norsufentanil by GC- or LC-MS