Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nortanshinone | 97399-70-7 | 1 MG
SDP

Catalog No. 5000418856
Encompass_Preferred

Nortanshinone is a pigment isolated from Dan-Shen, appearing as a brown to reddish-brown solid. It is classified under quinones and naphthalene quinones.

  • For laboratory research
  • Purity of 98.0%
  • Molecular formula: C17H12O4
  • Molecular weight: 280.27
  • Isolated from plants such as Leguminosae and Labiatae

Catalog No. 50-004-18856 Supplier Medchemexpress LLC Supplier No. HYN69141MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nortanshinone is a pigment isolated from Dan-Shen, appearing as a brown to reddish-brown solid. It is classified under quinones and naphthalene quinones.

  • For laboratory research
  • Purity of 98.0%
  • Molecular formula: C17H12O4
  • Molecular weight: 280.27
  • Isolated from plants such as Leguminosae and Labiatae

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.