Learn More
Medchemexpress LLC Nortanshinone | 97399-70-7 | 5 MG
SDP
Catalog No. 5000417301
Encompass_Preferred
Nortanshinone is a pigment isolated from Dan-Shen, intended for research use only. It has a molecular weight of 280.27 and a chemical formula of C17H12O4. This compound typically appears as a solid, ranging from brown to reddish brown.
  • High purity of 98.0%
  • Soluble in DMSO at 1.25 mg/mL (4.46 mM)
  • Isolated from plants such as Leguminosae and Labiatae
  • Classified as a quinone and naphthalene quinone
Catalog No. 50-004-17301 Supplier Medchemexpress LLC Supplier No. HYN69145MG
Nortanshinone is a pigment isolated from Dan-Shen, intended for research use only. It has a molecular weight of 280.27 and a chemical formula of C17H12O4. This compound typically appears as a solid, ranging from brown to reddish brown.
  • High purity of 98.0%
  • Soluble in DMSO at 1.25 mg/mL (4.46 mM)
  • Isolated from plants such as Leguminosae and Labiatae
  • Classified as a quinone and naphthalene quinone