Learn More
Guardian Corporation NORTECH 55 GAL STEEL DRUM/BUNG
Catalog No. NC1050875
Encompass
Durable, heavy-duty steel drum is ideal for capturing and storing wet or dry vacuumed materials. Epoxy/phenolic coated interior surface is resistant to chipping or peeling and provides corrosion protection. Circular ribs maintain structural integrity. Bottom bunghole allows for liquid drainage. Dimensions (in) 23.5 D x 33.5
Catalog No. NC1050875 Supplier Guardian Corporation Supplier No. N650
Durable, heavy-duty steel drum is ideal for capturing and storing wet or dry vacuumed materials. Epoxy/phenolic coated interior surface is resistant to chipping or peeling and provides corrosion protection. Circular ribs maintain structural integrity. Bottom bunghole allows for liquid drainage. Dimensions (in) 23.5 D x 33.5