Learn More
Honeywell™ North™ A59 Series Hard Hat
Catalog No. 19160221
Change view
Click to view available options
A59 Series Hard Hat
Catalog No.
19-160-221 A59 Series Hard Hat
1 options
This item is not returnable. View return policy
Catalog No. 19-160-221 Supplier Honeywell™ Supplier No. A59010000
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
A versatile and dependable design, with grooved HDPE outer shell, rain trough accessory slots, and 4-point plastic suspension. White hat with pin lock adjustment with 3-level height adjustment.
Meets ANSI Z89.1-2009 Type 1 Class E.
TRUSTED_SUSTAINABILITY
Specifications
Suspension Type 4-Point Pin-Lock