Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Adhesive Fingertip Bandages

Catalog No. 17986415
Change view
Click to view available options
Quantity:
40 Pk.
Product Type:
Fingertip Bandage
Color:
Beige
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Color
17-986-415 40 Pk. Fingertip Bandage Beige
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-986-415 Supplier Honeywell™ Supplier No. 010813

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fabric stretches with every movement

  • Designed with a strong adhesive to adhere securely and protect the wound
  • Island dressing provides a secure seal around the wound to help prevent contamination

TRUSTED_SUSTAINABILITY

Specifications

Product Type Fingertip Bandage
For Use With (Application) For effective first-aid treatment, wide range of wound care applications
Adhesive Aggressive Adhesive
Color Beige
Material Woven Fabric
Quantity 40 Pk.
Size Regular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.