Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Adhesive Knuckle Bandages

Catalog No. 17986411
Change view
Click to view available options
Quantity:
40/Box
Color:
Blue
Material:
Woven Fabric
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color Material
17-986-411 40/Box Blue Woven Fabric
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-986-411 Supplier Honeywell™ Supplier No. 017500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Greater protection for wound care applications

  • Conformable, fabric dressings stretch with every movement
  • Unique open weave fabric allows skin to breathe for fast healing
  • Aggressive adhesive sticks in grease, oil or water

TRUSTED_SUSTAINABILITY

Specifications

Product Type Knuckle Bandage
For Use With (Application) For visual detection only, excellent for use in restaurants and food service areas
Adhesive Aggressive Adhesive
Color Blue
Material Woven Fabric
Quantity 40/Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.