Learn More
Honeywell™ North™ Airline Respirator Assemblies
Catalog No. 19039649A
Change view
Click to view available options
7600 Series; Size Small; Includes 760008A w/CF2008, Welding Attachment
Catalog No.
19-039-649A 7600 Series; Size Small; Includes 760008A w/CF2008, Welding Attachment
1 options
Catalog No. 19-039-649A Supplier Honeywell™ Supplier No. CF2401SW
Converts the 5500 and 7700 Series half masks, and the 5400 and 7600 Series full facepieces into a continuous flow supplied air respirator.
Supplied air assembly with 7600 Series full facepiece with breathing tube and adapter, available in Small or Medium/Large.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Continuous Flow Airline Assembly
Model CF2401SW