Learn More
Honeywell™ North™ Airline Respirator Assemblies
Catalog No. 19039649B
Change view
Click to view available options
7600 Series; Size Medium/Large; Includes 760008A w/CF2007, Welding Attachment
Catalog No.
19-039-649B 7600 Series; Size Medium/Large; Includes 760008A w/CF2007, Welding Attachment
1 options
Catalog No. 19-039-649B Supplier Honeywell™ Supplier No. CF2401W
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Converts the 5500 and 7700 Series half masks, and the 5400 and 7600 Series full facepieces into a continuous flow supplied air respirator.
Supplied air assembly with 7600 Series full facepiece with breathing tube and adapter, available in Small or Medium/Large.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Continuous Flow Airline Assembly
Model CF2401W