Learn More
Honeywell™ North™ Airline Respirator Assemblies
Catalog No. 19039647A
Change view
Click to view available options
5500 Series; Size Small; Includes 550030 w/CF2007
Catalog No.
19-039-647A 5500 Series; Size Small; Includes 550030 w/CF2007
1 options
Catalog No. 19-039-647A Supplier Honeywell™ Supplier No. CF2101S
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Converts the 5500 and 7700 Series half masks, and the 5400 and 7600 Series full facepieces into a continuous flow supplied air respirator.
Supplied air assembly with 7600 Series full facepiece with breathing tube and adapter, available in Small or Medium/Large.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Continuous Flow Airline Assembly
Model CF2101S