Learn More
Honeywell™ North™ Airline Respirator Assemblies
Catalog No. 19039650C
Change view
Click to view available options
7700 Series; Size Large
Catalog No.
19-039-650C 7700 Series; Size Large
1 options
Catalog No. 19-039-650C Supplier Honeywell™ Supplier No. CF2201L
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Converts the 5500 and 7700 Series half masks, and the 5400 and 7600 Series full facepieces into a continuous flow supplied air respirator.
Supplied air assembly with 7600 Series full facepiece with breathing tube and adapter, available in Small or Medium/Large.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Continuous Flow Airline Assembly
Model CF2201L