Learn More
Honeywell™ North™ Aneroid Sphygmomanometer Adult Blood Pressure Cuff
Catalog No. 19310201
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-310-201 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. 19-310-201 Supplier Honeywell™ Supplier No. 320115G
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Refill item for North First Responder kits
  • Index and range markings on cuff
  • No-pin stop manometer
  • Finger/thumb air release valve
  • Gauge holder
TRUSTED_SUSTAINABILITY
Specifications
Product Type Aneroid Sphygmomanometer
Color Black or Blue
Includes Black or blue cuff, gauge holder, manometer (gauge), finger or thumb air release valve
Pressure 300 mmHg
Quantity 1 Ea.
Size Adult