Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Berman Airway Kit

Catalog No. 19310203
Change view
Click to view available options
Product Type:
Bermen Airway Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-310-203 Bermen Airway Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-310-203 Supplier Honeywell™ Supplier No. 423786KT
This item is not returnable. View return policy

Prevents patient from biting or swallowing their tongue during CPR

  • Contains six different sizes that range from 50 to 100mm (2 to 4 in.)
  • Material: Polyethylene
  • Color-coded by size
TRUSTED_SUSTAINABILITY

Specifications

Product Type Bermen Airway Kit
Material Polyethylene
Includes 6 Individually wrapped units
Quantity 1 Ea.
Size 50 to 100 mm (2 to 4 in.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.