Learn More
Honeywell™ North™ BP1000 Series Backpack Air-Purifying Respirator
Catalog No. 19040889A
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-040-889A Small
19-040-889B Medium
19-040-889C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-040-889A Supplier Honeywell™ Supplier No. BP1201S
This item is not returnable. View return policy
Convert your half mask or full facepiece to a welding respirator with this welding attachment. Puts cartridges on the back, increasing service life and minimizing weight to the facepiece.
  • Ideal for welding and other high-heat operations
  • Meets NIOSH 42 CFR Part 84 standards
Convenient backpack design allows cartridges and filters to be located on the user's back and out of the work zone—ideal for environments with high heat or sparks, such as welding, coke and smelting operations. Simply attach the backpack to North 7700 half mask cartridge connectors, then fasten the appropriate cartridges or filters to the backpack. No modifications, tools, or extra parts are needed.
Complete assembly includes backpack adapter and half-mask. Backpack adapter can also be purchased separately (below) to convert existing North facepieces to BP1000 Series air-purifying respirators
TRUSTED_SUSTAINABILITY
Specifications
Certifications (NIOSH) NIOSH 42 CFR Part 84
Size Small
For Use With (Application) Welding and other high-heat operations
For Use With (Equipment) North™ 5500, 7700, 5400 and 7600 Series facepieces
Includes Backpack adapter
Model BP1201S