Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Bulk First Aid Kits

Catalog No. 191302483
Change view
Click to view available options
Material:
Plastic
Sufficient For:
10 Persons
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Sufficient For
19-130-2483 Plastic 10 Persons
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-130-2483 Supplier Honeywell™ Supplier No. 0197004500F

missing translation for 'validMainframeMsg'

Designed for large facilities

  • Contain economical bulk supplies that can be readily accessed when needed
  • Kits are ideal for manufacturing settings where a wide variety of first aid items are required
  • Latex-free
  • Weatherproof
  • Supplies housed in waterproof metal or plastic case

Order Info

Rapid refill kits available separately.

Compliance

ANSI™ Z308.1-2003 compliant

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Manufacturing settings where a wide variety of first aid items are required
Product Type First Aid Kit
Certifications/Compliance ANSI Z308.1-2003
Material Plastic
Sufficient For 10 Persons
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.