Learn More
MSC North Bump Cap, Pinlock Adjustment, Vented, 4-Point Suspension, White, HDPE
Catalog No. 5000508689
Encompass
This vented bump cap offers lightweight, low-hazard head protection, ideal for various industrial settings. It features a pinlock adjustment and a 4-point suspension for a secure and comfortable fit. It is suitable for environments where head protection from minor bumps is needed, but should not be used to protect against falling objects.
  • Low hazard head protection
  • Well ventilated for comfort
  • Lightweight design
  • Pinlock adjustment for a secure fit
  • 4-point suspension for stability
  • Made from durable HDPE material
  • Suitable for food industry, mechanics, and truck drivers
  • White color for easy identification
  • Designed for environments not requiring ANSI compliant head protection
Catalog No. 50-005-08689 Supplier MSC Supplier No. 64994304
This vented bump cap offers lightweight, low-hazard head protection, ideal for various industrial settings. It features a pinlock adjustment and a 4-point suspension for a secure and comfortable fit. It is suitable for environments where head protection from minor bumps is needed, but should not be used to protect against falling objects.
  • Low hazard head protection
  • Well ventilated for comfort
  • Lightweight design
  • Pinlock adjustment for a secure fit
  • 4-point suspension for stability
  • Made from durable HDPE material
  • Suitable for food industry, mechanics, and truck drivers
  • White color for easy identification
  • Designed for environments not requiring ANSI compliant head protection