Learn More
Honeywell™ North™ Calcium Gluconate Gel
Catalog No. 19070469
Change view
Click to view available options
Volume (Metric):
25 g
Catalog No. Volume (Metric)
19-070-469 25 g
1 options
Catalog No. 19-070-469 Supplier Honeywell™ Supplier No. 28063068
Specifications
Product Type Calcium Gluconate Gel
For Use With (Application) Treatment of hydrofluoric acid burns, counteract an overdose of Magnesium Sulfate, treat hypocalcemia
Quantity 1 Ea.
Volume (English) 0.055 lb.
Volume (Metric) 25 g