Learn More
MSC North Coupling Assembly, Hansen, 1/2 in ID, for CF1000/CF2000 Series
Catalog No. 5000508312
Encompass
The North Coupling Assembly is a Hansen-style quick connect assembly designed to connect a breathing tube to a 1/2 in ID hose. It includes both male and female couplers and is approved for use with CF1000 and CF2000 Series supplied-air and powered air-purifying respirators. This assembly meets NIOSH and OSHA standards.
  • Includes one male plug and one female coupler
  • Connects breathing tube to 1/2 in ID hose
  • Designed for SAR systems
  • Hansen connection type
  • Meets NIOSH and OSHA standards
  • Compatible with AH9000 hoses
Catalog No. 50-005-08312 Supplier MSC Supplier No. 48464895
The North Coupling Assembly is a Hansen-style quick connect assembly designed to connect a breathing tube to a 1/2 in ID hose. It includes both male and female couplers and is approved for use with CF1000 and CF2000 Series supplied-air and powered air-purifying respirators. This assembly meets NIOSH and OSHA standards.
  • Includes one male plug and one female coupler
  • Connects breathing tube to 1/2 in ID hose
  • Designed for SAR systems
  • Hansen connection type
  • Meets NIOSH and OSHA standards
  • Compatible with AH9000 hoses