Learn More
MSC North Cut, Puncture & Abrasion-Resistant Gloves, Cut A3, Puncture 5, Abrasion 5, X-Small
Catalog No. 5000508745
Encompass
These gloves are engineered for robust protection against cuts, punctures, and abrasions in demanding work environments. Constructed with a lightweight 13-gauge design from HPPE and Dyneema, they provide advanced protection while ensuring wearer comfort and dexterity.
  • ANSI/ISEA Cut Resistance Level A3
  • ANSI/ISEA Puncture Resistance Level 5
  • ANSI/ISEA Abrasion Resistance Level 5
  • Size X-Small (numeric size 6)
  • 13 gauge construction
  • Primary material is HPPE
  • Polyurethane-coated for improved tactile sensitivity and digital dexterity
  • Seamless shell minimizes chafing
  • Stretchy and form-fitting for comfort
  • Polyurethane grip surface
  • Dyneema back and cuff material
  • Unisex design
Catalog No. 50-005-08745 Supplier MSC Supplier No. 40454936
These gloves are engineered for robust protection against cuts, punctures, and abrasions in demanding work environments. Constructed with a lightweight 13-gauge design from HPPE and Dyneema, they provide advanced protection while ensuring wearer comfort and dexterity.
  • ANSI/ISEA Cut Resistance Level A3
  • ANSI/ISEA Puncture Resistance Level 5
  • ANSI/ISEA Abrasion Resistance Level 5
  • Size X-Small (numeric size 6)
  • 13 gauge construction
  • Primary material is HPPE
  • Polyurethane-coated for improved tactile sensitivity and digital dexterity
  • Seamless shell minimizes chafing
  • Stretchy and form-fitting for comfort
  • Polyurethane grip surface
  • Dyneema back and cuff material
  • Unisex design