Learn More
MSC North Earmuffs, 25 dB, Three Position, Foam Cushion
Catalog No. 5000508500
Encompass
North Earmuffs are designed with a 25 dB Noise Reduction Rating. They feature a three-position band and foam cushions, making them suitable for environments requiring hearing protection.
  • 25 dB Noise Reduction Rating
  • Three-position band
  • Foam cushion material
  • Cushioned headband
  • Black cup color
  • Plastic cup material
  • Adjustable headband
  • Country of Origin: China
Catalog No. 50-005-08500 Supplier MSC Supplier No. 12227054
North Earmuffs are designed with a 25 dB Noise Reduction Rating. They feature a three-position band and foam cushions, making them suitable for environments requiring hearing protection.
  • 25 dB Noise Reduction Rating
  • Three-position band
  • Foam cushion material
  • Cushioned headband
  • Black cup color
  • Plastic cup material
  • Adjustable headband
  • Country of Origin: China