Learn More
Honeywell™ North™ First Aid Kit Refills
Catalog No. 17986192
Change view
Click to view available options
Product Type:
Forceps and Scissors
Catalog No. Product Type
17-986-192 Forceps and Scissors
1 options
This item is not returnable. View return policy
Catalog No. 17-986-192 Supplier Honeywell™ Supplier No. 020550
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Use these refill items to restock Fisher Scientific™ first aid kits and unitized North first aid kits
Rugged, reverse-tuck, color-coded packs permit quick identification. Clear plastic packaging lets color code show through, protects the pack from moisture, and includes a tear flap for quick opening.
  • Yellow: bandages
  • Blue: antiseptics
  • Green: special items such as insect sting or poison plant treatment, or miscellaneous items
  • Orange: personal protection (for products such as gloves)
Compliance
Each pack is stamped with lot number and expiration date (if applicable), in compliance with FDA requirements, and shows graphic instructions to assist in proper use of the item.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Unitized first aid kits
Product Type Forceps and Scissors
Certifications/Compliance FDA